SP Wilmad-LabGlass Dewar Traps, SP Industries
Supplier: SP Industries
ML-1825-702 ML-1825-700
14205-066EA 361.14 USD
14205-066 14206-552
SP Wilmad-LabGlass Dewar Traps, SP Industries
These borosilicate glass traps are similar to a Dewar flask, but feature the addition of a Standard Taper inner joint at bottom, and an outer joint of the same size and style extending in an upward direction from the side of the jacket near the top
The inside diameter of flask is approximately 32mm, and the inside height is approximately 120mm.