Anti-IgG(H+L) Goat Antibody (APC (Allophycocyanin))

Supplier: Southern Biotechnology
6061-11
100245-010EA 568.85 USD
100245-010
Anti-IgG(H+L) Goat Antibody (APC (Allophycocyanin))
Antibodies
Goat Anti-Hamster IgG(H+L)-APC antibody with minimal reactivity to mouse and rat proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR