Anti-Ly-6G/Ly-6C Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: RB6-8C5]

Supplier: Southern Biotechnology
1900-09 1900-09L
100244-052EA 238 USD
100244-052 100244-054
Anti-Ly-6G/Ly-6C Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: RB6-8C5]
Antibodies
Ly-6G/Ly-6C, formerly known as the myeloid differentiation antigen Gr-1, is a GPI-anchored protein. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation. It is also transiently expressed on monocytes in the bone marrow. In the periphery, Ly-6G/Ly-6C is expressed predominantly on neutrophils.

Type: Primary
Antigen: Ly-6G/Ly-6C
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR