Anti-I-Ek Mouse Monoclonal Antibody (Biotin) [clone: 17-3-3]

Supplier: Southern Biotechnology
1898-08
100244-042EA 324.31 USD
100244-042
Anti-I-Ek Mouse Monoclonal Antibody (Biotin) [clone: 17-3-3]
Antibodies
The monoclonal antibody 17-3-3 reacts with the I-Ek class II alloantigen on cells from mice of the H-2r haplotype. Reactivity with H-2d and H-2p haplotypes has not been observed. Cells from mice of the H-2b, H-2f, H-2g, H-2q, and H-2s haplotypes do not express I-E antigen.

Type: Primary
Antigen: I-Ek
Clonality: Monoclonal
Clone: 17-3-3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR