Anti-I-Ek Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 14-4-4S]

Supplier: Southern Biotechnology
1897-09
100244-036EA 223.62 USD
100244-036
Anti-I-Ek Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 14-4-4S]
Antibodies
The monoclonal antibody 14-4-4S reacts with the I-Ek class II alloantigen on cells from mice of the H-2d, H-2p, and H-2r haplotypes. Cells from mice of the H-2b, H-2f, H-2q, and H-2s haplotypes do not express I-E antigen. The antibody has been reported to cross-react with the rat MHC class II alloantigen RT1D.

Type: Primary
Antigen: I-Ek
Clonality: Monoclonal
Clone: 14-4-4S
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR