Anti-MHC Class II Rat Monoclonal Antibody (Biotin) [clone: NIMR-4]

Supplier: Southern Biotechnology
1895-08
100244-014EA 367.46 USD
100244-014
Anti-MHC Class II Rat Monoclonal Antibody (Biotin) [clone: NIMR-4]
Antibodies
The monoclonal antibody NIMR-4 reacts specifically with a non-polymorphic I-A-encoded epitope on MHC Class II antigens. Class II antigens are predominantly expressed on antigen-presenting cells including B lymphocytes, macrophages, dendritic cells, and certain epithelial cells.

Type: Primary
Antigen: MHC Class II
Clonality: Monoclonal
Clone: NIMR-4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR