Anti-LPAM-1 Rat Monoclonal Antibody (Biotin) [clone: DATK32]

Supplier: Southern Biotechnology
1870-08
100243-978EA 374 USD
100243-978
Anti-LPAM-1 Rat Monoclonal Antibody (Biotin) [clone: DATK32]
Antibodies
The lymphocyte Peyer’s patch adhesion molecule (LPAM-1), also known as integrin α4β7, is a member of the integrin family of cell surface receptors. It is expressed primarily on mucosal lymphocytes but is also present on NK cells and eosinophils. The α4β7 heterodimer mediates the binding of lymphocytes to its ligand, mucosal vascular addressin (MAdCAM-1) on the high endothelial venules, thereby directing the homing of lymphocytes into Peyer’s patches and intestinal lamina propria. The monoclonal antibody DATK32 recognizes a conformational epitope on the heterodimer.

Type: Primary
Antigen: LPAM-1
Clonality: Monoclonal
Clone: DATK32
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR