Anti-CD105 Rat Monoclonal Antibody [clone: MJ7/18]

Supplier: Southern Biotechnology
1860-01
100243-966EA 313.85 USD
100243-966
Anti-CD105 Rat Monoclonal Antibody [clone: MJ7/18]
Antibodies
CD105, also known as endoglin, is a disulfide-linked type II transmembrane homodimer that is predominantly expressed on vascular endothelial cells. It is also found on pre-erythroblasts, macrophages, and lymphoid and myeloid leukemic cells. Both mouse and human CD105 exhibit >70% sequence homology in their cytoplasmic domains with type III transforming growth factor β (TGFβ) receptor. CD105 has been shown to be one of several receptors for the various isoforms of TGFβ which in turn is one of a family of proteins involved in regulation of cell differentiation, migration of cells, and control of the immune response.

Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR