Anti-CD104 Rat Monoclonal Antibody [clone: 346-11A]

Supplier: Southern Biotechnology
1855-01
100243-962EA 313.85 USD
100243-962
Anti-CD104 Rat Monoclonal Antibody [clone: 346-11A]
Antibodies
CD104 represents the integrin β4 subunit that associates non-covalently with integrin α6 (CD49f) to form the α6β4 integrin heterodimer. The α6β4 complex is expressed on the basal surface of a variety of epithelial cell types particularly on stratified squamous epithelia. It is also found in peripheral nerves, in certain subsets of endothelial cells, and on immature thymocytes. It also has been found to be associated with a number of malignant tissues. Hemidesmosomal α6β4 plays an important role in the adhesion of epithelial cells to basement membranes via interactions with laminin and/or kalinin anchoring filaments. It is not known if the monoclonal antibody 346-11A can block this adhesion.

Type: Primary
Antigen: CD104
Clonality: Monoclonal
Clone: 346-11A
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR