Anti-CD79b Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HM79-12]

Supplier: Southern Biotechnology
1830-09
100243-958EA 258.92 USD
100243-958
Anti-CD79b Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: HM79-12]
Antibodies
Murine CD79 is a disulfide-linked heterodimer composed of an α chain (CD79a) and a β chain (CD79b) that associates non-covalently with membrane immunoglobulin (Ig) to form the B cell receptor (BCR) complex. Its expression is restricted to B lymphocytes first appearing on the surface at the pro-B cell stage prior to productive Ig gene rearrangements and remaining through all stages of B-cell differentiation prior to plasma cells. It has been proposed that the CD79 complex on pro-B cell surfaces may function to induce early B-cell differentiation.

Type: Primary
Antigen: CD79b
Clonality: Monoclonal
Clone: HM79-12
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR