Anti-CD81 Hamster Monoclonal Antibody [clone: 2F7]

Supplier: Southern Biotechnology
1825-14
100243-950EA 381.85 USD
100243-950
Anti-CD81 Hamster Monoclonal Antibody [clone: 2F7]
Antibodies
CD81, also known as TAPA-1, is an integral membrane protein expressed on a variety of cell types and has a high degree of sequence homology between human and mouse. CD81 is expressed on thymic stromal cells where it plays an important role in the transition of γδ+ T cells to more mature T cells with αβ T cell receptors. Immunohistochemical staining has revealed that its expression is localized to the subcapsular region of the thymus and, specifically, on cells that have distinct clustering patterns. It has been speculated that the ligand for CD81 is the pre-T cell receptor which is composed of a TCR β chain and glycoprotein pTα. The monoclonal antibody 2F7 can block thymocyte interaction with CD81 in vitro.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 2F7
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR