Anti-NK1.1 Mouse Monoclonal Antibody [clone: PK136]

Supplier: Southern Biotechnology
1805-14
100243-870EA 367.46 USD
100243-870
Anti-NK1.1 Mouse Monoclonal Antibody [clone: PK136]
Antibodies
NK1.1, a member of the NKR-P1 family of cell surface receptors, is a type II integral membrane glycoprotein with a C-type lectin domain. It is expressed as a disulfide-linked homodimer on all NK cells as well as subsets of thymocytes and peripheral T lymphocytes in selected strains of mice (e.g., C57BL/6, NZB, and CE). NK1.1 mediates cellular activation and differentiation, and is thought to have a particular role in generating Th2 cells. This product does not react with NK cells of BALB/c mice.

Type: Primary
Antigen: NK1.1
Clonality: Monoclonal
Clone: PK136
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR