Anti-Ly-49I Mouse Monoclonal Antibody [clone: YLI-90]

Supplier: Southern Biotechnology
1802-01
100243-832EA 309.92 USD
100243-832
Anti-Ly-49I Mouse Monoclonal Antibody [clone: YLI-90]
Antibodies
The monoclonal antibody YLI-90 reacts with the B6 allele of Ly49I, a member of the lectin-like homodimeric Ly49 family of cell surface receptors. Mouse Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells. The YLI-90 antibody stains ~45% of NK1.1+CD3- splenocytes from C57BL/6 mice.

Type: Primary
Antigen: Ly-49I
Clonality: Monoclonal
Clone: YLI-90
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR