Anti-CD152 Hamster Monoclonal Antibody [clone: 1B8]

Supplier: Southern Biotechnology
1790-14
100243-784EA 353.08 USD
100243-784
Anti-CD152 Hamster Monoclonal Antibody [clone: 1B8]
Antibodies
The lymphocyte surface antigen CD152, also known as CTLA-4, is related to the costimulatory molecule CD28 and both molecules share common B7 family counter-receptors. However CD152 is thought to be a negative regulator of T cell activation and may play a role in apoptotic control of T cells. CD152 is relatively conserved among humans, mice, and chickens.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: 1B8
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR