Anti-TCRbeta Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: H57-597]

Supplier: Southern Biotechnology
1785-09
100243-766EA 166.08 USD
100243-766
Anti-TCRbeta Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: H57-597]
Antibodies
The αβ TCR is expressed on T lymphocytes of all mouse strains tested. The H57-597 monoclonal antibody does not react with γδ TCR-bearing cells. Plate-bound or soluble H57-597 activates αβ TCR-bearing T cells. In vitro and in vivo, H57-597 can induce immature thymocytes to undergo apoptosis. In vivo administration of H57-597 has been shown to deplete αβ TCR-bearing cells to near completion and prevent graft rejection.

Type: Primary
Antigen: TCRbeta
Clonality: Monoclonal
Clone: H57-597
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR