Anti-TCRgammadelta Hamster Monoclonal Antibody [clone: UC7-13D5]

Supplier: Southern Biotechnology
1780-14
100243-756EA 396.23 USD
100243-756
Anti-TCRgammadelta Hamster Monoclonal Antibody [clone: UC7-13D5]
Antibodies
The TCRγδ heterodimer of the CD3/T cell receptor complex is clonotypic and consists of Ig-like variable and constant domains. It is expressed during thymopoiesis and on the small subpopulation of γδ TCR-expressing T lymphocytes in the periphery. The majority of T cells present in some epithelial tissues are γδ+ and have limited receptor diversity. The TCRγδ heterodimer recognizes peptide antigen bound to MHC antigens and subsequent signal transduction mediated by the invariant chains leads to T cell activation. The monoclonal antibody UC7-13D5 is specific for the γδ heterodimer and plate-bound UC7-13D5 activates γδ TCR-bearing cells. This antibody does not react with αβ TCR-expressing T cells.

Type: Primary
Antigen: TCRgammadelta
Clonality: Monoclonal
Clone: UC7-13D5
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR