Anti-BP-1 Rat Monoclonal Antibody [clone: FG35.4]

Supplier: Southern Biotechnology
1770-01
100243-730EA 277.23 USD
100243-730
Anti-BP-1 Rat Monoclonal Antibody [clone: FG35.4]
Antibodies
BP-1, also known as Ly-51 and 6C3 antigen, is a type II homodimeric transmembrane glycoprotein formed by two disulfide-linked chains of 140 kDa. Originally identified on the surface of pre-B cell lymphomas the cDNA encoding BP-1 has been cloned and found to represent aminopeptidase A. It is expressed at high levels on bone marrow stromal cell lines which support in vitro B lymphopoiesis and on a wide variety of mouse tissues known to possess aminopeptidase activity. Subsets of normal bone marrow pre-B and B cells express low levels of BP-1 which is rapidly upregulated on pre-B cells in the presence of IL-7. The BP-1 marker is commonly used to identify B cells in different stages of development.

Type: Primary
Antigen: BP-1
Clonality: Monoclonal
Clone: FG35.4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR