Anti-Ly-6C Rat Monoclonal Antibody [clone: HK1.4]

Supplier: Southern Biotechnology
1760-01
100243-722EA 285.08 USD
100243-722
Anti-Ly-6C Rat Monoclonal Antibody [clone: HK1.4]
Antibodies
Ly-6C is a member of the Ly-6 multigene family of type V glycophosphatidylinositol-anchored cell surface proteins. It is expressed on bone marrow cells, monocytes/macrophages, neutrophils, endothelial cells, and T-cell subsets. Mice with the Ly-6.2 allotype (e.g., AKR, C57BL, C57BR, C57L, DBA/2, PL, SJL, SWR, 129) have subsets of CD4+Ly-6C+ and CD8+Ly-6C+ cells while Ly-6.1 strains (e.g., A, BALB/c, CBA, C3H/He, DBA/1, NZB) have only CD8+Ly-6C+ lymphocytes. Ly-6C may play a role in the development and maturation of lymphocytes.

Type: Primary
Antigen: Ly-6C
Clonality: Monoclonal
Clone: HK1.4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR