Anti-CD90.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]

Supplier: Southern Biotechnology
1750-11
100243-698EA 320.38 USD
100243-698
Anti-CD90.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Antibodies
CD90 (Thy-1) is a GPI-anchored molecule and one of the smallest members of the immunoglobulin superfamily consisting of a single V-set domain. The Thy-1.2 alloantigen is expressed on all thymocytes, peripheral T lymphocytes, and some intraepithelial T cells of most mouse strains. It is also expressed in the brain and at varying levels on other non-lymphoid tissues. The 30-H12 monoclonal antibody does not cross react with mouse and rat strains bearing the Thy-1.1 alloantigen (e.g., AKR/J, PL).

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR