Anti-CD90 Rat Monoclonal Antibody [clone: G7]

Supplier: Southern Biotechnology
1740-14
100243-684EA 353.08 USD
100243-684
Anti-CD90 Rat Monoclonal Antibody [clone: G7]
Antibodies
CD90 (Thy-1), a GPI-anchored molecule and one of the smallest members of the immunoglobulin superfamily of cell surface receptors, consists of a single V-set domain. It is expressed on thymocytes, peripheral T lymphocytes, some intraepithelial T lymphocytes, and neurons of all mouse strains. The monoclonal antibody G7 stimulates T-cell proliferation and IL-2 secretion via signaling through the T-cell receptor/CD3 complex. G7 has also been reported to promote apoptosis of thymocytes and CTL clones and to mediate adhesion of thymocytes to thymic stroma.

Type: Primary
Antigen: CD90
Clonality: Monoclonal
Clone: G7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR