Anti-CD80 Rat Monoclonal Antibody [clone: 1G10]

Supplier: Southern Biotechnology
1730-01
100243-640EA 277.23 USD
100243-640
Anti-CD80 Rat Monoclonal Antibody [clone: 1G10]
Antibodies
CD80 (B7-1) is a type I transmembrane glycoprotein and a member of the immunoglobulin superfamily of cell surface receptors. It is constitutively expressed by dendritic cells, monocytes, and peritoneal macrophages. Expression of CD80 on B cells is upregulated after activation by various means including stimulation by LPS, IL-4, and surface Ig receptor crosslinking with peak expression occurring between 48 and 72 hours post-activation. The interaction of CD80 with its ligands, CD28 and CD152 (CTLA-4), provides a potent costimulatory signal for T cells activated through the CD3/T cell receptor complex. The monoclonal antibody 1G10 blocks binding of CTLA-4 to CD80 but it does not block stimulation of T cells by natural antigen-presenting cells.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR