Anti-CD71 Rat Monoclonal Antibody [clone: RI7217]

Supplier: Southern Biotechnology
1720-14
100243-630EA 381.85 USD
100243-630
Anti-CD71 Rat Monoclonal Antibody [clone: RI7217]
Antibodies
CD71, a type II transmembrane glycoprotein expressed on the cell surface as a homodimer, is the transferrin receptor. It is essential for the growth of normal and neoplastic cells. It is expressed at low levels on resting B and T lymphocytes but is upregulated during responses to antigens and mitogens presumably reflecting the iron dependence of proliferation. Its expression declines with maturation and differentiation. CD71 plays a critical role in cell proliferation by controlling the supply of iron which is essential for many metabolic pathways through the binding and endocytosis of transferrin, the major iron-carrying protein. It may also be involved in signal transduction via its association with the T cell receptor ζ chain. The monoclonal antibody RI7217 inhibits cell proliferation in vitro.

Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: RI7217
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR