Anti-CD69 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H1.2F3]

Supplier: Southern Biotechnology
1715-02
100243-608EA 353.08 USD
100243-608
Anti-CD69 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H1.2F3]
Antibodies
CD69, also known as very early activation (VEA) antigen, is a disulfide-linked transmembrane homodimer whose differentially glycosylated subunits range from 35-39 kDa. It is a C-type lectin most closely related to the NKR-P1 and Ly-49 NK cell-activation molecules. CD69 is widely expressed on hematopoietic cells, including lymphocytes, neutrophils and eosinophils. Although not detectable on resting lymphocytes, its expression is rapidly upregulated upon activation of T, B and NK cells, and neutrophils. Constitutive expression of CD69 on subsets of thymocytes suggests that it may be involved in regulation of developmental events in addition to its role in activation of a variety of hematopoietic cells. The monoclonal antibody H1.2F3 augments PMA-induced T-cell proliferation and induces redirected lysis of Fc receptor-bearing target cells by NK cells.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR