Anti-CD54 Rat Monoclonal Antibody [clone: YN1/1.7.4]

Supplier: Southern Biotechnology
1701-14
100243-586EA 387.08 USD
100243-586
Anti-CD54 Rat Monoclonal Antibody [clone: YN1/1.7.4]
Antibodies
CD54, a member of the immunoglobulin superfamily of cell surface receptors and also known as Intercellular Adhesion Molecule-1 (ICAM-1), is a type I transmembrane glycoprotein that mediates important cell/cell interactions. CD54 is expressed on endothelial cells, dendritic cells, keratinocytes, and lymphocytes. Its expression is upregulated by inflammatory mediators such as IFN γ, IL-1, TNF-α, lipopolysaccharide, and phorbol esters. Endothelial CD54 contributes to the extravasation of leukocytes from blood vessels, particularly in areas of inflammation. CD54 on antigen-presenting cells contributes to antigen-specific T-cell activation, presumably by enhancing interactions between T cell and antigen-presenting cells. These adhesion reactions are mediated via the binding of CD54 to its major ligands, LFA-1 and Mac-1.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: YN1/1.7.4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR