Anti-CD49e Rat Monoclonal Antibody [clone: 5H10.27]

Supplier: Southern Biotechnology
1695-14
100243-576EA 367.46 USD
100243-576
Anti-CD49e Rat Monoclonal Antibody [clone: 5H10.27]
Antibodies
The monoclonal antibody 5H10.27 reacts with the α5 chain (VLA-5) of the CD49e/CD29 heterodimeric fibronectin receptor. It is expressed on thymocytes, activated T cells, mast cells, and a variety of mouse cell lines. Soluble 5H10.27 antibody inhibits VLA-5-mediated functions in vitro. Immobilized 5H10.27 has also been demonstrated to costimulate the proliferative response of CD8+ T cells to plate-bound anti-CD3 monoclonal antibody.

Type: Primary
Antigen: CD49e
Clonality: Monoclonal
Clone: 5H10.27
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR