Anti-CD45RC Rat Monoclonal Antibody (Biotin) [clone: GL24]

Supplier: Southern Biotechnology
1685-08
100243-558EA 402.77 USD
100243-558
Anti-CD45RC Rat Monoclonal Antibody (Biotin) [clone: GL24]
Antibodies
CD45RC represents an exon C-dependent isoform of CD45 glycoprotein. It is expressed at high density on all B lymphocytes, at intermediate density on most CD8+ splenic T cells, and on two subpopulations of CD4+ splenic T cells with high and low density of expression. CD45RC is also present on a small subset of thymocytes.

Type: Primary
Antigen: CD45RC
Clonality: Monoclonal
Clone: GL24
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR