Anti-CD45R Rat Monoclonal Antibody (Cy5®) [clone: RA3-6B2]

Supplier: Southern Biotechnology
1665-15
100243-540EA 359.62 USD
100243-540
Anti-CD45R Rat Monoclonal Antibody (Cy5®) [clone: RA3-6B2]
Antibodies
CD45R, also known as B220, a member of the protein tyrosine phosphate family and a major cell surface glycoprotein, represents a restricted form of the CD45 family which primarily recognizes only cells of B-lineage from pro-B cell through mature B lymphocytes and prior to the availability of anti-CD19 monoclonal antibodies was commonly used as a pan B-cell marker. It also reacts with certain activated T cells as well as non-MHC-restricted lytically active lymphokine-activated killer (LAK) cells. In vivo administration of RA3-6B2 has been shown to affect differentiation of both T and B cells in normal mice and reduce the level of anti-DNA antibodies and lymphadenopathies in MRL/lpr mice.

Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human, Feline
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR