Anti-CD45 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: I3/2.3]

Supplier: Southern Biotechnology
1660-11
100243-514EA 258.92 USD
100243-514
Anti-CD45 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: I3/2.3]
Antibodies
CD45 is the common leukocyte antigen and is expressed on all cells of hematopoietic origin except erythrocytes. CD45 exists as a number of different isoforms which result from alternative RNA splicing of exons 4, 5 and 6. The I3/2.3 monoclonal antibody recognizes a framework epitope present on all CD45 isoforms. The CD45 molecule has been implicated as playing a key role in thymic development and in antigen-driven proliferation of T cells.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: I3/2.3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR