Anti-CD40 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1C10]

Supplier: Southern Biotechnology
1645-02
100243-480EA 381.85 USD
100243-480
Anti-CD40 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1C10]
Antibodies
CD40 is a type I cell surface protein belonging to the tumor necrosis factor superfamily of cell surface receptors. In mice it is expressed on B lineage cells, follicular dendritic cells, thymic epithelium, and interdigitating cells in the T-cell zone of secondary lymphoid organs. CD40 first becomes detectable on a subset of small pre-B II cells in bone marrow with the levels of CD40 expression increasing thereafter during B cell maturation. Immature B cells (IgM+IgDloB220lo) express intermediate levels of CD40, whereas mature B cells (IgM+IgDhiB220hi) express high levels. CD40 has a central role in B cell growth and differentiation, and signaling through CD40 in combination with IL-4 reportedly induces immunoglobulin isotype switching and secretion of IgE. The agonistic 1C10 antibody closely resembles gp39/CD40 ligand in its ability to stimulate proliferation of small, resting B lymphocytes in the absence of other cofactors.

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 1C10
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR