Anti-CD30 Hamster Monoclonal Antibody (Biotin) [clone: mCD30.1]

Supplier: Southern Biotechnology
1620-08
100243-406EA 367.46 USD
100243-406
Anti-CD30 Hamster Monoclonal Antibody (Biotin) [clone: mCD30.1]
Antibodies
CD30 is a type I transmembrane protein and a member of the tumor necrosis factor receptor superfamily of proteins. Murine CD30 is expressed predominantly in the thymus and it is inducible in mouse splenocytes stimulated with pokeweed mitogen or concanavalin A. In anti-CD3ε-activated spleen cells, CD30 is expressed primarily on the surface of CD8+ T cells with peak expression on days 4 and 5. Stimulation of CD30+ CTL lines with plate-bound anti-CD30 directly signals IL-5 but not IFN-γ production. While these studies demonstrate that CD30 directs cytokine secretion and suggest that CD30 may play a pivotal role in the pattern of cytokine production by T cells, the precise roles of CD30 and its ligand (CD153) in T-cell development have not been clearly defined.

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: mCD30.1
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR