Anti-CD28 Hamster Monoclonal Antibody (Biotin) [clone: PV-1]

Supplier: Southern Biotechnology
1615-08
100243-390EA 324.31 USD
100243-390
Anti-CD28 Hamster Monoclonal Antibody (Biotin) [clone: PV-1]
Antibodies
CD28 is a type I disulfide-linked homodimer that is constitutively expressed on most thymocytes, at low density on nearly all CD4+ and CD8+ peripheral T lymphocytes and at very low levels on NK cells. Its expression is upregulated upon T-cell activation. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) on B cells and other antigen presenting cells and plays an important role in the interaction between T cells and B cells. CD28 is a costimulatory receptor involved in many, but not all, T-cell independent immune responses.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: PV-1
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR