Anti-CD25 Rat Monoclonal Antibody [clone: 3C7]

Supplier: Southern Biotechnology
1600-01
100243-354EA 277.23 USD
100243-354
Anti-CD25 Rat Monoclonal Antibody [clone: 3C7]
Antibodies
The IL-2 receptor (IL-2R) exists in three alternative forms made up from the individual components of CD25, CD122, and CD132. CD25 represents the low affinity α chain of the IL-2R, a type I transmembrane glycoprotein containing two CCP domains. It is rich in O-linked carbohydrates and has a short cytoplasmic tail. CD25 is expressed on activated T cells, B cells, NK cells, and monocytes of all mouse strains tested. Expression of CD25 on activated T lymphocytes is transitory and endogenously regulated. CD25 is expressed on precursor B cells in bone marrow. Its expression is initiated by functional rearrangement and expression of IgM heavy chain genes and is down-regulated when immature B cells mature and express IgD. It is expressed at a higher level on CD4+CD8+ thymocytes. It is also expressed on cultured epidermal Langerhans cells. The biochemical features of murine CD25 have been characterized in detail. The 3C7 monoclonal antibody reacts with an epitope of CD25 which is distinct from that recognized by clone 7D4. 3C7, used in combination with 7D4 in culture, results in higher levels of inhibition of proliferation driven by IL-2 and generation of alloreactive CTL than either of these monoclonal antibodies alone.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: 3C7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR