Anti-CD25 Rat Monoclonal Antibody [clone: 7D4]

Supplier: Southern Biotechnology
1595-01
100243-336EA 324.31 USD
100243-336
Anti-CD25 Rat Monoclonal Antibody [clone: 7D4]
Antibodies
The IL-2 receptor (IL-2R) exists in three alternative forms made up from the individual components of CD25, CD122, and CD132. CD25 represents the low affinity α chain of the IL-2R, a type I transmembrane glycoprotein containing two CCP domains. It is rich in O-linked carbohydrates and has a short cytoplasmic tail. CD25 is expressed on activated T cells, B cells, NK cells, and monocytes of all mouse strains tested. It is transiently expressed at a low level during B cell development in bone marrow on the CD45R/B220low TdT- sIg- pre-B II and CD45R/B220low TdT- sIgM+sIgD- immature B cells, but not on the CD45R/B220low TdT+ sIg- pro-B/pre-B I stage nor on CD45R/B220high TdT- sIgM+sIgD+ mature B cells. CD25 is expressed at a higher level during very early T-cell development in fetal and adult thymus. The 7D4 monoclonal antibody reacts with an epitope between amino acids 125 and 212 that is distinct from that recognized by the monoclonal antibody 3C7 (SB Cat. No. 1600). While the monoclonal antibody 7D4 does not block the binding of IL-2 to CD25, when used in combination with the monoclonal antibody 3C7 it results in higher levels of inhibition of IL-2-driven proliferation than either of these two monoclonal antibodies alone.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: 7D4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR