Anti-CD24 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 91]

Supplier: Southern Biotechnology
1590-02
100243-326EA 353.08 USD
100243-326
Anti-CD24 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 91]
Antibodies
CD24, also known as heat stable antigen, is a phosphatidylinositol-linked cell surface glycoprotein that is expressed at all stages of B cell development and on most thymocytes. The absence of expression from mature T cells is closely associated with their maturation from CD4+CD8+CD24+ thymocytes to either CD4+CD8-CD24- or CD4-CD8+CD24- T cells. CD24 is also present on mouse granulocytes, monocytes, Langerhans cells and erythrocytes. Its ligand is P-selectin (CD62P). CD24 may play a role in regulation of B cell proliferation and differentiation, and it has been proposed that heat stable antigen promotes homotypic adhesion between B lymphocytes.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: 91
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR