Anti-CD18 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: C71/16]

Supplier: Southern Biotechnology
1570-09
100243-254EA 209.23 USD
100243-254
Anti-CD18 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: C71/16]
Antibodies
CD18 represents the common β2 integrin subunit that associates non-covalently with the α/β chains of CD11a, CD11b, and CD11c to form various integrin heterodimers. It is expressed strongly on lymphocytes and monocytes and weakly on granulocytes. CD18 mediates a variety of heterotypic and homotypic intercellular adhesion reactions and it regulates the ligand-binding activates of the various CD11/CD18 complexes. The monoclonal antibody C71/16 has not been reported to inhibit cell adhesion.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: C71/16
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR