Anti-CD11a Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: I21/7]

Supplier: Southern Biotechnology
1555-09L 1555-09
100243-198EA 273.31 USD
100243-198 100243-200
Anti-CD11a Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: I21/7]
Antibodies
The monoclonal antibody I21/7 reacts with the α subunit of mouse and human CD11a, also known as LFA-1, and a member of the integrin family of cell adhesion molecules. The antigen is expressed on thymocytes, T and B lymphocytes, and on non-lymphoid cells in the bone marrow. CD11a appears to be an essential component for T-cell activation by antigen-presenting cells and is important for cell-mediated killing. Costimulatory signaling by LFA-1 is achieved by pairing with its ligand, CD54/ICAM-1.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: I21/7
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR