Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: 43449]

Supplier: Southern Biotechnology
1525-08
100243-050EA 320.38 USD
100243-050
Anti-CD2 Rat Monoclonal Antibody (Biotin) [clone: 43449]
Antibodies
CD2, also known as leukocyte function-associated antigen-2 (LFA-2), is a member of the immunoglobulin superfamily of cell adhesion molecules. It is expressed on the surface of >95% of thymocytes, and essentially all B cells, T cells and NK cells. In the mouse, CD2 is the receptor for CD48. CD2 is thought to play a major role in mediating T-cell adhesion, T-cell recognition/activation, T-cell signaling, and intrathymic differentiation.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 43449
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR