Anti-CD49d Rat Monoclonal Antibody [clone: PS/2]

Supplier: Southern Biotechnology
1520-14
100243-042EA 324.31 USD
100243-042
Anti-CD49d Rat Monoclonal Antibody [clone: PS/2]
Antibodies
VLA-4 belongs to the β1-integrin family, which functions both as cell-matrix and cell-cell receptors. As a matrix receptor VLA-4 binds to an alternatively spliced domain of fibronectin, while as a cellular receptor it associates with its counter-receptor ligand VCAM-1. In addition to CD44/Pgp-1, VLA-4 has been shown to be important for lymphopoiesis, and is also believed to be important for extravasation of lymphocytes into sites of inflammation as a second signal in addition to LFA/ICAM-1. CD49d is expressed in low amounts on resting lymphocytes and monocytes, is absent or minimally detectable on stromal cells, whereas virtually all lymphoid and myeloid cells in bone marrow and in long-term culture are positive. Antibody PS/2 also reacts with human VLA-4.

Type: Primary
Antigen: CD49d
Clonality: Monoclonal
Clone: PS/2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR