Anti-Fibrillin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11C1.3]

Supplier: Southern Biotechnology
1405-02
100242-998EA 525.69 USD
100242-998
Anti-Fibrillin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 11C1.3]
Antibodies
The fibrillins are extracellular matrix glycoproteins which occur as a major component of a subset of connective tissue microfibrils. These microfibrils have a beaded appearance and a cross-sectional diameter of 10-12 nm. In elastic tissues, these structures are thought to provide the scaffold onto which elastin is assembled to form elastic fibers, although their function in non-elastic tissues is unclear. There is an aberrant expression of fibrillin in Marfan syndrome. The epitope for the monoclonal antibody 11C1.3 has been found to be within exons 18-20 (a.a. 723-909).

Type: Primary
Antigen: Fibrillin
Clonality: Monoclonal
Clone: 11C1.3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Japanese Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR