Anti-IL-6 Rat Monoclonal Antibody (Biotin) [clone: MP5-32C11]

Supplier: Southern Biotechnology
10208-08
100242-410EA 330.85 USD
100242-410
Anti-IL-6 Rat Monoclonal Antibody (Biotin) [clone: MP5-32C11]
Antibodies
Biotin Anti-Mouse IL-6 antibody for use in immunohistochemistry / immunocytochemistry, ELISA, ELISpot, and multiplex assays.

Type: Primary
Antigen: IL-6
Clonality: Monoclonal
Clone: MP5-32C11
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR