Anti-IL-2 Rat Monoclonal Antibody (Biotin) [clone: JES6-5H4]

Supplier: Southern Biotechnology
10202-08
100242-392EA 330.85 USD
100242-392
Anti-IL-2 Rat Monoclonal Antibody (Biotin) [clone: JES6-5H4]
Antibodies
Biotin Anti-Mouse IL-2 antibody for use in flow cytometry, immunohistochemistry, immunoprecipitation, ELISA, ELISpot, and multiplex assays.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR