Anti-IL-2 Rat Monoclonal Antibody [clone: JES6-5H4]

Supplier: Southern Biotechnology
10202-01
100242-390EA 287.69 USD
100242-390
Anti-IL-2 Rat Monoclonal Antibody [clone: JES6-5H4]
Antibodies
Purified Anti-Mouse IL-2 antibody for use in flow cytometry, immunohistochemistry, immunoprecipitation, ELISA, and ELISpot assays.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR