Anti-IgM Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
1020-09 1020-09S
100242-378EA 381.85 USD
100242-378 100242-380
Anti-IgM Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Mouse IgM-PE antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR