Anti-IgM Goat Antibody (Texas Red®)

Supplier: Southern Biotechnology
1020-07
100242-374EA 209.23 USD
100242-374
Anti-IgM Goat Antibody (Texas Red®)
Antibodies
Goat Anti-Mouse IgM-Texas Red antibody with minimal reactivity to human proteins for use in flow cytometry and immunohistochemistry / immunocytochemistry assays.

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR