Anti-Ig Goat Antibody (Cy5®)

Supplier: Southern Biotechnology
1012-15
100242-338EA 281.15 USD
100242-338
Anti-Ig Goat Antibody (Cy5®)
Antibodies
Goat F(ab')2 Anti-Mouse Ig-CY5 antibody with minimal reactivity to human proteins for use in flow cytometry, immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR