Anti-Ig Goat Antibody (Spectral Red® RPE (R-Phycoerythrin)/Cy5®)

Supplier: Southern Biotechnology
1012-13
100242-336EA 367.46 USD
100242-336
Anti-Ig Goat Antibody (Spectral Red® RPE (R-Phycoerythrin)/Cy5®)
Antibodies
Goat F(ab')2 Anti-Mouse Ig-PE/CY5 (SPRD) antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Spectral Red® RPE (R-Phycoerythrin)/Cy5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR