Anti-IL-4 Rat Monoclonal Antibody (Biotin) [clone: MP4-25D2]

Supplier: Southern Biotechnology
10117-08
100242-308EA 330.85 USD
100242-308
Anti-IL-4 Rat Monoclonal Antibody (Biotin) [clone: MP4-25D2]
Antibodies
Biotin Anti-Human IL-4 antibody for use in flow cytometry, immunohistochemistry, ELISA, ELISpot, and multiplex assays.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Rhesus, Cynomolgus, Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR