Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]

Supplier: Southern Biotechnology
10117-01
100242-306EA 287.69 USD
100242-306
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]
Antibodies
Purified Anti-Human IL-4 antibody for use in flow cytometry, immunohistochemistry, ELISA, and ELISpot assays.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Rhesus, Cynomolgus, Baboon
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR