Anti-IL-3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]

Supplier: Southern Biotechnology
10116-08
100242-304EA 396.23 USD
100242-304
Anti-IL-3 Rat Monoclonal Antibody (Biotin) [clone: BVD3-1F9]
Antibodies
Biotin Anti-Human IL-3 antibody for use in flow cytometry, immunohistochemistry, ELISA, western blotting, and immunoprecipitation assays.

Type: Primary
Antigen: IL-3
Clonality: Monoclonal
Clone: BVD3-1F9
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR