Anti-Ig Goat Antibody (Biotin)

Supplier: Southern Biotechnology
1010-08
100242-278EA 172.62 USD
100242-278
Anti-Ig Goat Antibody (Biotin)
Antibodies
Goat Anti-Mouse Ig-Biotin antibody with minimal reactivity to human proteins for use in flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR